Anti-1N Tau Rabbit Monoclonal Antibody [clone: EPR25206-82]

Lieferant: Abcam
AB314889-20UL AB314889-100UL AB314889-1ML
ABCAAB314889-20ULEA 236 CHF
ABCAAB314889-20UL ABCAAB314889-100UL ABCAAB314889-1ML
Anti-1N Tau Rabbit Monoclonal Antibody [clone: EPR25206-82]
Antikörper
Rabbit monoclonal [EPR25206-82] to 1N Tau.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: 1N Tau
Clonality: Monoclonal
Clone: EPR25206-82
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR