Anti-PKC gamma Rabbit Monoclonal Antibody [clone: EPR28643-68]
Lieferant: Abcam
AB317315-20UL AB317315-100UL AB317315-1ML
ABCAAB317315-20ULEA 236 CHF
ABCAAB317315-20UL ABCAAB317315-100UL ABCAAB317315-1ML
Anti-PKC gamma Rabbit Monoclonal Antibody [clone: EPR28643-68]
Antikörper
Rabbit monoclonal [EPR28643-68] to PKC gamma.
Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: PKC gamma
Clonality: Monoclonal
Clone: EPR28643-68
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat