Anti-Somatostatin 28 Rabbit Recombinant Multiclonal Antibody [clone: RM1087]
Lieferant: Abcam
AB315106-20UL AB315106-100UL AB315106-1ML
ABCAAB315106-20ULEA 200 CHF
ABCAAB315106-20UL ABCAAB315106-100UL ABCAAB315106-1ML
Anti-Somatostatin 28 Rabbit Recombinant Multiclonal Antibody [clone: RM1087]
Antikörper
Rabbit recombinant multiclonal [RM1087] to Somatostatin 28.
Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Somatostatin 28
Clonality: Recombinant Multiclonal
Clone: RM1087
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat