Anti-RIM1 Rabbit Monoclonal Antibody [clone: EPR29131-30]

Lieferant: Abcam
AB317450-20UL AB317450-100UL AB317450-1ML
ABCAAB317450-20ULEA 236 CHF
ABCAAB317450-20UL ABCAAB317450-100UL ABCAAB317450-1ML
Anti-RIM1 Rabbit Monoclonal Antibody [clone: EPR29131-30]
Antikörper
Rabbit monoclonal [EPR29131-30] to RIM1.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: RIM1
Clonality: Monoclonal
Clone: EPR29131-30
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR