Anti-Semaphorin 4D/CD100 Rabbit Monoclonal Antibody [clone: EPR26247-28]
Lieferant: Abcam
AB320634-20UL AB320634-100UL AB320634-1ML
ABCAAB320634-20ULEA 236 CHF
ABCAAB320634-20UL ABCAAB320634-100UL ABCAAB320634-1ML
Anti-Semaphorin 4D/CD100 Rabbit Monoclonal Antibody [clone: EPR26247-28]
Antikörper
Rabbit monoclonal [EPR26247-28] to Semaphorin 4D/CD100.
Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Semaphorin 4D/CD100
Clonality: Monoclonal
Clone: EPR26247-28
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse