Anti-Lactate Dehydrogenase B/LDH-B Rabbit Monoclonal Antibody [clone: EP1565Y] (Alexa Fluor® 647)

Lieferant: Abcam
AB320697-10UL AB320697-100UL
ABCAAB320697-10ULEA 230 CHF
ABCAAB320697-10UL ABCAAB320697-100UL
Anti-Lactate Dehydrogenase B/LDH-B Rabbit Monoclonal Antibody [clone: EP1565Y] (Alexa Fluor® 647)
Antikörper
Alexa Fluor® 647 Rabbit monoclonal [EP1565Y] to Lactate Dehydrogenase B/LDH-B.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Lactate Dehydrogenase B/LDH-B
Clonality: Monoclonal
Clone: EP1565Y
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR