Anti-Glucose Transporter GLUT1 Rabbit Monoclonal Antibody [clone: EPR3915] (FITC (Fluorescein Isothiocyanate))
Lieferant: Abcam
AB322306-10UL AB322306-100UL
ABCAAB322306-10ULEA 230 CHF
ABCAAB322306-10UL ABCAAB322306-100UL
Anti-Glucose Transporter GLUT1 Rabbit Monoclonal Antibody [clone: EPR3915] (FITC (Fluorescein Isothiocyanate))
Antikörper
FITC Rabbit monoclonal [EPR3915] to Glucose Transporter GLUT1.
Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Glucose Transporter GLUT1
Clonality: Monoclonal
Clone: EPR3915
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human