Anti-H-FABP Rabbit Monoclonal Antibody [clone: EPR22017-264]

Lieferant: Abcam
AB281064-100 AB281064-1
ABCAAB281064-100EA 841 CHF
ABCAAB281064-100 ABCAAB281064-1
Anti-H-FABP Rabbit Monoclonal Antibody [clone: EPR22017-264]
Antikörper
Rabbit monoclonal [EPR22017-264] to H-FABP - BSA and Azide free (Detector).

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: H-FABP
Clonality: Monoclonal
Clone: EPR22017-264
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR