Anti-Eph receptor A1/EphA1 Rabbit Monoclonal Antibody [clone: EPR18738-119]

Lieferant: Abcam
AB242822-1 AB242822-100
ABCAAB242822-1EA 9020 CHF
ABCAAB242822-1 ABCAAB242822-100
Anti-Eph receptor A1/EphA1 Rabbit Monoclonal Antibody [clone: EPR18738-119]
Antikörper
Rabbit monoclonal [EPR18738-119] to Eph receptor A1/EphA1 - BSA and Azide free (Detector).

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Eph receptor A1/EphA1
Clonality: Monoclonal
Clone: EPR18738-119
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR