Anti-Thioredoxin / TRX Rabbit Monoclonal Antibody [clone: EPR23843-213]
Lieferant: Abcam
AB284974-100 AB284974-1
ABCAAB284974-100EA 908 CHF
ABCAAB284974-100 ABCAAB284974-1
Anti-Thioredoxin / TRX Rabbit Monoclonal Antibody [clone: EPR23843-213]
Rabbit monoclonal [EPR23843-213] to Thioredoxin / TRX - BSA and Azide free (Capture).
Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Thioredoxin / TRX
Clonality: Monoclonal
Clone: EPR23843-213
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse