Anti-SUZ12 Rabbit Monoclonal Antibody [clone: EPR26230-82]

Lieferant: Abcam
AB307891-100 AB307891-20UL AB307891-1ML
ABCAAB307891-100EA 672 CHF
ABCAAB307891-100 ABCAAB307891-20UL ABCAAB307891-1ML
Anti-SUZ12 Rabbit Monoclonal Antibody [clone: EPR26230-82]
Antikörper
Rabbit monoclonal [EPR26230-82] to SUZ12.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: SUZ12
Clonality: Monoclonal
Clone: EPR26230-82
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR