Anti-KMT5A / SETD8 / Pr-SET7 Rabbit Monoclonal Antibody [clone: EPR24381-24]

Lieferant: Abcam
AB300517-100 AB300517-20UL AB300517-1ML
ABCAAB300517-100EA 684 CHF
ABCAAB300517-100 ABCAAB300517-20UL ABCAAB300517-1ML
Anti-KMT5A / SETD8 / Pr-SET7 Rabbit Monoclonal Antibody [clone: EPR24381-24]
Antikörper
Rabbit monoclonal [EPR24381-24] to KMT5A / SETD8 / Pr-SET7.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: KMT5A / SETD8 / Pr-SET7
Clonality: Monoclonal
Clone: EPR24381-24
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR