Anti-NTF2 Mouse Monoclonal Antibody [clone: 5A3]

Lieferant: Abcam
AB254146-100 AB254146-1MG
ABCAAB254146-100EA 739 CHF
ABCAAB254146-100 ABCAAB254146-1MG
Anti-NTF2 Mouse Monoclonal Antibody [clone: 5A3]
Antikörper
Mouse monoclonal [5A3] to NTF2.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: NTF2
Clonality: Monoclonal
Clone: 5A3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR