Anti-PHIP Rabbit Monoclonal Antibody [clone: EPR27150-82]
Lieferant: Abcam
AB309963-100UG AB309963-1MG
ABCAAB309963-100UGEA 987 CHF
ABCAAB309963-100UG ABCAAB309963-1MG
Anti-PHIP Rabbit Monoclonal Antibody [clone: EPR27150-82]
Antikörper
Rabbit monoclonal [EPR27150-82] to PHIP - BSA and Azide free.
Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: PHIP
Clonality: Monoclonal
Clone: EPR27150-82
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human