Anti-6X His tag® Rabbit Monoclonal Antibody [clone: EPR20547]

Lieferant: Abcam
AB213204-100 AB213204-1ML
ABCAAB213204-100EA 460 CHF
ABCAAB213204-100 ABCAAB213204-1ML
Anti-6X His tag® Rabbit Monoclonal Antibody [clone: EPR20547]
Antikörper
Rabbit monoclonal [EPR20547] to 6X His tag® - ChIP Grade.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: 6X His tag®
Clonality: Monoclonal
Clone: EPR20547
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Recombinant full length protein
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR