Anti-MIP-1 alpha/CCL3 Rabbit Monoclonal Antibody [clone: EPR19900-147]

Lieferant: Abcam
AB245251-100 AB245251-1ML
ABCAAB245251-100EA 733 CHF
ABCAAB245251-100 ABCAAB245251-1ML
Anti-MIP-1 alpha/CCL3 Rabbit Monoclonal Antibody [clone: EPR19900-147]
Antikörper
Rabbit monoclonal [EPR19900-147] to MIP-1 alpha/CCL3.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: MIP-1 alpha/CCL3
Clonality: Monoclonal
Clone: EPR19900-147
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR