Anti-TLS/FUS Mouse Monoclonal Antibody [clone: Z-FUS-5]

Lieferant: Abcam
AB283297-100 AB283297-1
ABCAAB283297-100EA 987 CHF
ABCAAB283297-100 ABCAAB283297-1
Anti-TLS/FUS Mouse Monoclonal Antibody [clone: Z-FUS-5]
Antikörper
Mouse monoclonal [Z-FUS-5] to TLS/FUS - Chimeric – BSA and Azide free.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TLS/FUS
Clonality: Monoclonal
Clone: Z-FUS-5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR