Anti-PCNA Mouse Monoclonal Antibody [clone: 24/PCNA]

Lieferant: Abcam
AB280093-1 AB280093-100
ABCAAB280093-1EA 8870 CHF
ABCAAB280093-1 ABCAAB280093-100
Anti-PCNA Mouse Monoclonal Antibody [clone: 24/PCNA]
Antikörper
Mouse monoclonal [24/PCNA] to PCNA - BSA and Azide free.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PCNA
Clonality: Monoclonal
Clone: 24/PCNA
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR