Anti-RAB27A+RAB27B Rabbit Monoclonal Antibody [clone: EPR27120-68]
Lieferant: Abcam
AB313576-1MG AB313576-100UG
ABCAAB313576-1MGEA 8870 CHF
ABCAAB313576-1MG ABCAAB313576-100UG
Anti-RAB27A+RAB27B Rabbit Monoclonal Antibody [clone: EPR27120-68]
Antikörper
Rabbit monoclonal [EPR27120-68] to RAB27A + RAB27B - BSA and Azide free.
Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: RAB27A+RAB27B
Clonality: Monoclonal
Clone: EPR27120-68
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat