Anti-GPNMB Rabbit Monoclonal Antibody [clone: EPR22593-27]

Lieferant: Abcam
AB272302-100 AB272302-1
ABCAAB272302-100EA 841 CHF
ABCAAB272302-100 ABCAAB272302-1
Anti-GPNMB Rabbit Monoclonal Antibody [clone: EPR22593-27]
Antikörper
Rabbit monoclonal [EPR22593-27] to GPNMB - BSA and Azide free (Detector).

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: GPNMB
Clonality: Monoclonal
Clone: EPR22593-27
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR