Anti-Placental alkaline phosphatase (PLAP) Rabbit Monoclonal Antibody [clone: SP15]

Lieferant: Abcam
AB218848-1 AB218848-100
ABCAAB218848-1EA 8870 CHF
ABCAAB218848-1 ABCAAB218848-100
Anti-Placental alkaline phosphatase (PLAP) Rabbit Monoclonal Antibody [clone: SP15]
Antikörper
Rabbit monoclonal [SP15] to Placental alkaline phosphatase (PLAP) - BSA and Azide free.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Placental alkaline phosphatase (PLAP)
Clonality: Monoclonal
Clone: SP15
Conjugation:
Epitope: Not determined
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR