Anti-FAM160A2 Rabbit Monoclonal Antibody [clone: EPR13604]

Lieferant: Abcam
AB184160-100 AB184160-1ML
ABCAAB184160-100EA 678 CHF
ABCAAB184160-100 ABCAAB184160-1ML
Anti-FAM160A2 Rabbit Monoclonal Antibody [clone: EPR13604]
Antikörper
Rabbit monoclonal [EPR13604] to FAM160A2.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: FAM160A2
Clonality: Monoclonal
Clone: EPR13604
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR