Anti-CD38 Rabbit Monoclonal Antibody [clone: EPR4106]

Lieferant: Abcam
AB176886-100 AB176886-1
ABCAAB176886-100EA 987 CHF
ABCAAB176886-100 ABCAAB176886-1
Anti-CD38 Rabbit Monoclonal Antibody [clone: EPR4106]
Antikörper
Rabbit monoclonal [EPR4106] to CD38 - Low endotoxin, Azide free.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: EPR4106
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR