Anti-SLC16A3/MCT 4 Rabbit Monoclonal Antibody [clone: EPR28177-30]
Lieferant: Abcam
AB308529-1MG AB308529-100UG
ABCAAB308529-1MGEA 8870 CHF
ABCAAB308529-1MG ABCAAB308529-100UG
Anti-SLC16A3/MCT 4 Rabbit Monoclonal Antibody [clone: EPR28177-30]
Antikörper
Rabbit monoclonal [EPR28177-30] to SLC16A3/MCT 4 - BSA and Azide free.
Type: Primary
Antigen: SLC16A3/MCT 4
Clonality: Monoclonal
Clone: EPR28177-30
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat