Anti-HDAC4 + 5 + 9 Rabbit Monoclonal Antibody [clone: EPR19010-77]

Lieferant: Abcam
AB195241-100 AB195241-20UL AB195241-1ML
ABCAAB195241-100EA 690 CHF
ABCAAB195241-100 ABCAAB195241-20UL ABCAAB195241-1ML
Anti-HDAC4 + 5 + 9 Rabbit Monoclonal Antibody [clone: EPR19010-77]
Antikörper
Rabbit monoclonal [EPR19010-77] to HDAC4 + 5 + 9.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HDAC4 + 5 + 9
Clonality: Monoclonal
Clone: EPR19010-77
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR