Anti-Angiotensin Converting Enzyme 1 Rabbit Monoclonal Antibody [clone: EPR22291-42]
Lieferant: Abcam
AB242352-100 AB242352-1
ABCAAB242352-100EA 908 CHF
ABCAAB242352-100 ABCAAB242352-1
Anti-Angiotensin Converting Enzyme 1 Rabbit Monoclonal Antibody [clone: EPR22291-42]
Antikörper
Rabbit monoclonal [EPR22291-42] to Angiotensin Converting Enzyme 1 - BSA and Azide free (Detector).
Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Angiotensin Converting Enzyme 1
Clonality: Monoclonal
Clone: EPR22291-42
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human