Anti-Tau Rabbit Monoclonal Antibody [clone: EPR26001-28]

Lieferant: Abcam
AB312308-100UL AB312308-10UL AB312308-20UL AB312308-1ML
ABCAAB312308-100ULEA 672 CHF
ABCAAB312308-100UL ABCAAB312308-10UL ABCAAB312308-20UL ABCAAB312308-1ML
Anti-Tau Rabbit Monoclonal Antibody [clone: EPR26001-28]
Antikörper
Rabbit monoclonal [EPR26001-28] to Tau.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Tau
Clonality: Monoclonal
Clone: EPR26001-28
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR