Anti-FADS2 Rabbit Monoclonal Antibody [clone: EPR27357-82]

Lieferant: Abcam
AB314317-100UL AB314317-20UL AB314317-1ML
ABCAAB314317-100ULEA 672 CHF
ABCAAB314317-100UL ABCAAB314317-20UL ABCAAB314317-1ML
Anti-FADS2 Rabbit Monoclonal Antibody [clone: EPR27357-82]
Antikörper
Rabbit monoclonal [EPR27357-82] to FADS2.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: FADS2
Clonality: Monoclonal
Clone: EPR27357-82
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR