Anti-Dopamine Transporter Rabbit Monoclonal Antibody [clone: EPR19695]
Lieferant: Abcam
AB184451-100 AB184451-20UL AB184451-1ML
ABCAAB184451-100EA 763 CHF
ABCAAB184451-100 ABCAAB184451-20UL ABCAAB184451-1ML
Anti-Dopamine Transporter Rabbit Monoclonal Antibody [clone: EPR19695]
Antikörper
Rabbit monoclonal [EPR19695] to Dopamine Transporter.
Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Dopamine Transporter
Clonality: Monoclonal
Clone: EPR19695
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human