Anti-COL1A2 Rabbit Monoclonal Antibody [clone: EPR27415-30]
Lieferant: Abcam
AB308455-100 AB308455-20UL AB308455-1ML
ABCAAB308455-100EA 672 CHF
ABCAAB308455-100 ABCAAB308455-20UL ABCAAB308455-1ML
Anti-COL1A2 Rabbit Monoclonal Antibody [clone: EPR27415-30]
Antikörper
Rabbit monoclonal [EPR27415-30] to COL1A2.
Type: Primary
Antigen: COL1A2
Clonality: Monoclonal
Clone: EPR27415-30
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human