Anti-SMARCC1/BAF155 Rabbit Monoclonal Antibody [clone: EPR25109-77]
Lieferant: Abcam
AB305037-100 AB305037-20UL AB305037-1ML
ABCAAB305037-100EA 636 CHF
ABCAAB305037-100 ABCAAB305037-20UL ABCAAB305037-1ML
Anti-SMARCC1/BAF155 Rabbit Monoclonal Antibody [clone: EPR25109-77]
Antikörper
Rabbit monoclonal [EPR25109-77] to SMARCC1/BAF155.
Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SMARCC1/BAF155
Clonality: Monoclonal
Clone: EPR25109-77
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat