Anti-Progesterone Receptor Rabbit Monoclonal Antibody [clone: YR85] (Alexa Fluor® 488)

Lieferant: Abcam
AB313737-10UL AB313737-100UL
ABCAAB313737-10ULEA 230 CHF
ABCAAB313737-10UL ABCAAB313737-100UL
Anti-Progesterone Receptor Rabbit Monoclonal Antibody [clone: YR85] (Alexa Fluor® 488)
Antikörper
Alexa Fluor® 488 Rabbit monoclonal [YR85] to Progesterone Receptor.

Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Progesterone Receptor
Clonality: Monoclonal
Clone: YR85
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR