Anti-hSET1/SET1 Rabbit Monoclonal Antibody [clone: EPR23173-30]
Lieferant: Abcam
AB243639-100 AB243639-1ML
ABCAAB243639-100EA 739 CHF
ABCAAB243639-100 ABCAAB243639-1ML
Anti-hSET1/SET1 Rabbit Monoclonal Antibody [clone: EPR23173-30]
Antikörper
Rabbit monoclonal [EPR23173-30] to hSET1/SET1.
Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: hSET1/SET1
Clonality: Monoclonal
Clone: EPR23173-30
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human