Anti-hnRNP A1 (citrulline R92) Rabbit Monoclonal Antibody [clone: EPR20174]

Lieferant: Abcam
AB208026-100 AB208026-1ML
ABCAAB208026-100EA 648 CHF
ABCAAB208026-100 ABCAAB208026-1ML
Anti-hnRNP A1 (citrulline R92) Rabbit Monoclonal Antibody [clone: EPR20174]
Antikörper
Rabbit monoclonal [EPR20174] to hnRNP A1 (citrulline R92).

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: hnRNP A1
Clonality: Monoclonal
Clone: EPR20174
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR