Anti-Trans-Golgi network integral membrane protein 1 Rabbit Monoclonal Antibody [clone: EPR24831-36]
Lieferant: Abcam
AB283702-100 AB283702-1
ABCAAB283702-100EA 987 CHF
ABCAAB283702-100 ABCAAB283702-1
Anti-Trans-Golgi network integral membrane protein 1 Rabbit Monoclonal Antibody [clone: EPR24831-36]
Antikörper
Rabbit monoclonal [EPR24831-36] to Trans-Golgi network integral membrane protein 1 - BSA and Azide free.
Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Trans-Golgi network integral membrane protein 1
Clonality: Monoclonal
Clone: EPR24831-36
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse