Anti-Noradrenaline transporter Rabbit Monoclonal Antibody [clone: EPR23527-534]

Lieferant: Abcam
AB254361-100 AB254361-20UL AB254361-1ML
ABCAAB254361-100EA 690 CHF
ABCAAB254361-100 ABCAAB254361-20UL ABCAAB254361-1ML
Anti-Noradrenaline transporter Rabbit Monoclonal Antibody [clone: EPR23527-534]
Antikörper
Rabbit monoclonal [EPR23527-534] to Noradrenaline transporter.

Bitte beachten All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Noradrenaline transporter
Clonality: Monoclonal
Clone: EPR23527-534
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR