Molecular model: college organic set, Molymod®
Lieferant: Spiring Enterprises
Molymod®
SPIRMMS-051EA 0 CHF
SPIRMMS-051
Molecular model: college organic set, Molymod®
Modelle Molekularbiologie
The Molymod system is the unique dual-scale system of high quality molecular models. Atom parts are made of solid plastic spheres, available with from one to six holes in the usual angular orientations.
Atoms: 30 hydrogen atoms (white, molydome type), 14 carbon atoms (black, 4 hole, tetrahedral), 6 carbon atoms (black, 5 hole, trigonal bipyramidal), 4 nitrogen atoms (blue, 4 hole, tetrahedral), 6 oxygen atoms (red, 2 hole, angular), 8 halogen atoms (green, 1 hole), 2 bromine atoms (green, 1 hole), 2 iodine atoms (purple, 1 hole), 2 metal atoms (grey, 6 holes, octahedral).
Bonds: 40 medium links (grey, single bonds), 12 long flexible links (grey, double/triple bonds), 50 short links (white, all bond types).
One tool is used for disassembling models containing short link bonds.
Bestellinformation: Contains three link types to represent chemical bonds and flat shapes representing p-orbitals for illustrating orbital interactions.
Verpackung: In a sturdy plastic storage box.