Ammonium-1-pyrrolidincarbodithioat

Lieferant: Roth Carl

ROTH5486.1EA 61.75 CHF
ROTH5486.1
Ammonium-1-pyrrolidincarbodithioat
Ammonium-1-pyrrolidincarbodithioat
Formel: C₅H₁₂N₂S₂
MW: 146,3 · 18,0 g/mol
MDL Number: MFCD00012720
CAS-Nummer: 5108-96-3
EINECS: 225-834-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR