These culture tubes, made of PS, are an extension to the tissue culture flask product range.
- 3-in-1 product: cultivate, examine and spin cells in the same vessel
- Volume graduations up to the tip
- Conical form eases removal of pellet
- Centrifugation in a 15 or 50 ml standard tube adapter at 1200×g
Tissue Culture Flat Tube 10 cm2: Has 10 cm2 surface activated growth area with new surface quality resulting in crystal clear transparency. Tube has large opening for complete access of pipettes and scrappers. The sloped and flattened upper side helps in reduced refraction. It has great optical clarity for visual control with inverse microscopes.
Tissue Culture Tube 20 cm²: With 20 cm² surface activated growth area over a length of 50 mm. Tube has specially formed 10-sided comb from VENT screw cap with 'Click' avoids inadvertent rolling and enables regaining previous positions.
The Triple Bagged (3-B) packaging system meets the strict cleanroom standards wich prevents the contamination enters the highly sensitive area of production. Bags are security-opening style. The innermost bag 1 is labeled with the product details: product number, product description, lot number. Packaging is ideal for sterile transfer of products essential. Each product are packaged individually in a tear-open bag. Each bag is labelled with a security number: 3 = external, 2 = moderate, 1 = innermost bag. The safety-open-style peel-open bags are increasingly sized, so that during handling of one bag the next inner bag is not accidentally broken rendering it contaminated.
With the standard packaging, contamination adhering to the outside of the bag could be unintentionally carried into clean room areas. The 3-B packaging system prevents contamination from reaching highly sensitive production areas, thus significantly increasing safety for handling substances in a sterile environment.
Zertifizierungen: ISO 9001:2008 certified.
Bestellinformation: Optional tissue culture rack (fits 8× tubes in a horizontal position for storage in a CO2 incubator) must be ordered separately.
Verpackung: Culture tubes with 10 cm2 growth area are supplied as segmental packaging (4 tubes/bag) for scratch-free transportation. Bag slits with laser perforation for easy opening mechanism. And the culture tubes with 20 cm2 growth area are supplied in a convenient re-closable zipper bag with laser perforation.