Culture tubes with screw cap
Lieferant: TPP
91106 91243 391243
TPPA91106EA 588 CHF
TPPA91106 TPPA91243 TPPA391243
Culture tubes with screw cap
Röhrchen Kulturröhrchen
These culture tubes, made of PS, are an extension to the tissue culture flask product range.
  • 3-in-1 product: cultivate, examine and spin cells in the same vessel
  • Volume graduations up to the tip
  • Conical form eases removal of pellet
  • Centrifugation in a 15 or 50 ml standard tube adapter at 1200×g
Tissue Culture Flat Tube 10 cm²: Has 10 cm² surface activated growth area with new surface quality resulting in crystal clear transparency. Tube has large opening for complete access of pipettes and scrappers. The sloped and flattened upper side helps inMore Product Information