Cell culture flasks with reclosable lid
Lieferant: TPP
90552 90652
TPPA90552EA 313 CHF
TPPA90552 TPPA90652
Cell culture flasks with reclosable lid
Kolben Zellkulturkolben
Polystyrene tissue culture flasks with multi-use reclosable liquid-tight lid.
  • Surface activated, uniform flat growth surface
  • Crystal clear transparency
  • Geometry of the neck reduces risk of media slop over
  • Complete access to flask chamber from above
  • Stable and slip-free stacking of multiple flasks
Flasks are available only with filter screw caps. If a vented screw cap is used, a build-up of more than 0,03 bar inside the flask could press open the lid.
Zertifizierungen: ISO 9001:2008 certified.
Bestellinformation: Optional barrier which creates a growth area of 115 cm² can be ordered separately.