Blood collection tubes with clot activator, vacuum, Vacutest®
Lieferant: KIMA SAS DI CHIARIN RENZO
11020 11030 11002 11010 11005
720-2734EA 46.9 CHF
720-2734 720-2735 720-2730 720-2731 720-2763
Blood collection tubes with clot activator, vacuum, Vacutest®
Klinisches Diagnosezubehör Klinische Probenentnahme und Probenvorbereitung Entnahme und Vorbereitung von Blutproben
The test tubes are made from Polyethylene terephthalate (PET). Resistant to mechanical stress, centrifugation, breakage during standard procedures of use and in case of accidental falls. PET is a transparent material which enables the visual inspection of the blood and other biological liquids collected. Labels are made from self-adhesive paper for specific medical use, size 40×20 mm, printed in three colours.
  • Available in three sizes
  • Colours of the closure system are compliant with ISO 6710 and EN 14820 standards for the identification of the tubes
  • 18