Diphenyleniodoniumchlorid 97+%

Lieferant: Thermo Scientific
Achtung

ACRO328660250EA 192 CHF
ACRO328660250
Diphenyleniodoniumchlorid 97+%
Diphenyleneiodonium chloride
Formel: C₁₂H₈ClI
MW: 314,55 g/mol
Melting Pt: 312…322 °C
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00214165
CAS-Nummer: 4673-26-1

Order Now

Spezifikation Testergebnisse

Appearance Off-white Powder
TLC ≥97.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR