2-Amino-3-methoxypyrazin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Methoxypyrazin-2-amine

AS30649G1 AS30649G5
ACTIAS30649G1EA 48 CHF
ACTIAS30649G1 ACTIAS30649G5
2-Amino-3-methoxypyrazin
2-Amino-3-methoxypyrazine
Formel: C₅H₇N₃O
MW: 125,1300 g/mol
MDL Number: MFCD05722793
CAS-Nummer: 4774-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR