Luminol (3-Aminophthalhydrazid)
Lieferant: EUROCLONE
EMR165005
EMR165010
EMR165025
EURLEMR165005EA
0
CHF
EURLEMR165005
EURLEMR165010
EURLEMR165025
Luminol (3-Aminophthalhydrazid)
Luminol (3-Aminophthalhydrazid)
Formel:
C₈H₇N₃O₂ |
MDL Number:
MFCD00006890 CAS-Nummer: 521-31-3 EINECS: 208-309-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...