Diphenylborsäure(2-aminoethyl)ester (2-APB), Millipore®

Lieferant: Merck Millipore Calbiochem

100065.100EA 246 CHF
100065.100
Diphenylborsäure(2-aminoethyl)ester (2-APB), Millipore®
Diphenylborsäure(2-aminoethyl)ester (2-APB)
Formel: C₁₄H₁₆BNO
MW: 225,1 g/mol
MDL Number: MFCD00014823
CAS-Nummer: 524-95-8
EINECS: 208-366-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR