Benzo[k]fluoranthen
Lieferant: EHRENSTORFER GMBH
EHERC20580000EA
116
CHF
EHERC20580000
Benzo[k]fluoranthen
Benzo[k]fluoranthen
Formel:
C₂₀H₁₂ |
MDL Number:
MFCD00046287 CAS-Nummer: 207-08-9 EINECS: 205-916-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...