Riboflavin-5'-phosphat Natriumsalz Dihydrat

Lieferant: Molekula

MOLEM31244850EA 0 CHF
MOLEM31244850
Riboflavin-5'-phosphat Natriumsalz Dihydrat
Riboflavin-5'-phosphat Natriumsalz Dihydrat
Formel: C₁₇H₂₀N₄NaO₉P·2H₂O
MW: 514,36 g/mol (478,34 anhy)
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00150993
CAS-Nummer: 6184-17-4
EINECS: 204-988-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR