Magermilchpulver, Pulver

Lieferant: EUROCLONE

EURLEMR180001EA 0 CHF
EURLEMR180001 EURLEMR180500
Magermilchpulver, Pulver
Magermilchpulver
Lager Temperatur: Raumtemperatur
CAS-Nummer: 999999-99-4
EINECS: 310-127-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR