Cumolhydroperoxid
Lieferant: Cayman Chemical
CAYM703118-1EA
104
CHF
CAYM703118-1
Cumolhydroperoxid
Cumolhydroperoxid
Formel:
C₉H₁₂O₂ MW: 152,20 g/mol |
MDL Number:
MFCD00002129 CAS-Nummer: 80-15-9 EINECS: 201-254-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...